Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 2-3/8 X 2-3/4 inch Frame Style DelIVery Rectangle Sterile, 1624W, 100/Box
Catalog No. 502831837
Encompass_Preferred
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Adhesive is gentle on skin, and conformable dressing flexes with skin for patient comfort Picture-frame delivery makes accurate placement easy 2-3/8 x 2-3/4 inches
Catalog No. 50-283-1837 Supplier Med Vet International Supplier No. 235697BX
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Adhesive is gentle on skin, and conformable dressing flexes with skin for patient comfort Picture-frame delivery makes accurate placement easy 2-3/8 x 2-3/4 inches