Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 2-3/8 X 2-3/4 inch Frame Style DelIVery Rectangle Sterile, 1614, 400/Case
Catalog No. 502832269
Encompass_Preferred
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Reinforced notches reduce mechanical stress from heavy catheters Polyurethane film is thin for better contour over high-profile devices Picture-frame delivery makes accurate placement easy
Catalog No. 50-283-2269 Supplier Med Vet International Supplier No. 468778CS
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Reinforced notches reduce mechanical stress from heavy catheters Polyurethane film is thin for better contour over high-profile devices Picture-frame delivery makes accurate placement easy