Learn More
Med Vet International IV Dressing With Chg 3M Tegaderm Chg (Chlorhexidine Gluconate) / Film 3 1/2 X 4 1/2 inch Sterile, 1657, 100/Case
Catalog No. 502831899
Encompass_Preferred
Integrated I.V. dressing combines infection reduction with site visibility, catheter securement, and breathability Proven to reduce CRBSI and vascular catheter colonization Provides immediate and continuous antimicrobial protection with integrated chlorhexidine gluconate (CHG) gel pad Helps keep catheters in place with large securement tape strip and conforming keyhole notch Transparent film and gel pad allows continuous site visibility to easily assess for early signs of infection Breathable film coating provides a barrier to microbes and external contaminants White
Catalog No. 50-283-1899 Supplier Med Vet International Supplier No. 663782CS
Integrated I.V. dressing combines infection reduction with site visibility, catheter securement, and breathability Proven to reduce CRBSI and vascular catheter colonization Provides immediate and continuous antimicrobial protection with integrated chlorhexidine gluconate (CHG) gel pad Helps keep catheters in place with large securement tape strip and conforming keyhole notch Transparent film and gel pad allows continuous site visibility to easily assess for early signs of infection Breathable film coating provides a barrier to microbes and external contaminants White