Learn More
Med Vet International Absorbent Transparent Film Dressing 3M Tegaderm Absorbent 4-3/8 X 5 inch Oval Sterile, 90801, 10/Box
Catalog No. 502829965
Encompass_Preferred
Transparent film and absorbent pad allows wound or surgical incision to be monitored without dressing removal Clear design takes the guesswork out of application over the wound or surgical incision Novel acrylic polymer pad is breathable and designed to absorb low to moderate exudate to promote an optimal healing environment Dressing does not break down in the wound or surgical incision The non-bulky design helps to prevent dressing roll or lift due to friction from linens, clothing or prosthetics Protects from external contaminants, bodily fluids, bacteria, viruses and water ingress Wear time of up to 7 days gives patients extended protection beyond the initial hospital stay and into the home
Catalog No. 50-282-9965 Supplier Med Vet International Supplier No. 1220168BX
Transparent film and absorbent pad allows wound or surgical incision to be monitored without dressing removal Clear design takes the guesswork out of application over the wound or surgical incision Novel acrylic polymer pad is breathable and designed to absorb low to moderate exudate to promote an optimal healing environment Dressing does not break down in the wound or surgical incision The non-bulky design helps to prevent dressing roll or lift due to friction from linens, clothing or prosthetics Protects from external contaminants, bodily fluids, bacteria, viruses and water ingress Wear time of up to 7 days gives patients extended protection beyond the initial hospital stay and into the home