Learn More
Med Vet International Picc/Cvc Securement Device + Tegaderm IV Advanced Securement Dressing 3M Tegaderm Film 4 X 6-1/8 inch Sterile, 1839-2100, 80/Case
Catalog No. 502831482
Encompass_Preferred
Sutureless securement balances reliable adhesion with gentleness to skin Removes cleanly, without causing undue pain or distress Accommodates the majority of PICC and CVC catheters up to and including 12 Fr Designed to be worn for up to 7 days
Catalog No. 50-283-1482 Supplier Med Vet International Supplier No. 875305CS
Sutureless securement balances reliable adhesion with gentleness to skin Removes cleanly, without causing undue pain or distress Accommodates the majority of PICC and CVC catheters up to and including 12 Fr Designed to be worn for up to 7 days