Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 4 X 4-1/2 inch With Border Film Backing Acrylic Adhesive Oval Sterile, 90611, 10/Box
Catalog No. 502832419
Encompass_Preferred
The 3M Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering 3M's Tegaderm dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This dressing will not stick to wounds or disrupt healing tissue
Catalog No. 50-283-2419 Supplier Med Vet International Supplier No. 465141BX
The 3M Tegaderm high-performance foam adhesive dressing is suitable for use on wounds and ulcers, and with small medical devices It's easy to apply, thanks to its spoke delivery system The thin film backing blocks bacteria and viruses from entering 3M's Tegaderm dressing is made of soft, flexible foam to comfortably conform to your skin's contours Its multiple-layer construction helps it absorb fluids and keep wound areas dry This dressing will not stick to wounds or disrupt healing tissue