Learn More
Med Vet International Foam Dressing 3M Tegaderm 2 X 2 inch With Border Film Backing Silicone Adhesive Square Sterile, 90643, 60/Case
Catalog No. 502830250
Encompass_Preferred
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 2 x 2 inches
Catalog No. 50-283-0250 Supplier Med Vet International Supplier No. 1079670CS
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 2 x 2 inches