Learn More
Med Vet International Foam Dressing 3M Tegaderm High Performance 8 X 8 inch Without Border Film Backing NonAdhesive Square Sterile, 90603, 5/Box
Catalog No. 502831706
Encompass_Preferred
Multi-layer design provides high absorbency with high breathability to reduce the risk of maceration Adapts to changing levels of exudate to maintain moisture balance for optimal wound healing Demonstrates high fluid handling capacity under compression Dressing does not stick to wound bed thereby minimizing disruption of healing tissue Film backing prevents strike-through and protects against external bacteria and viruses
Catalog No. 50-283-1706 Supplier Med Vet International Supplier No. 465960BX
Multi-layer design provides high absorbency with high breathability to reduce the risk of maceration Adapts to changing levels of exudate to maintain moisture balance for optimal wound healing Demonstrates high fluid handling capacity under compression Dressing does not stick to wound bed thereby minimizing disruption of healing tissue Film backing prevents strike-through and protects against external bacteria and viruses