Learn More
Med Vet International Thin Hydrocolloid Dressing 3M Tegaderm Thin 4 X 4 inch Square, 90022, 100/Case
Catalog No. 502831883
Encompass_Preferred
Thin, absorbent and highly conformable May be used as a protective dressing on at-risk skin May be used on intact skin to protect from friction Film border on oval sizes seals to prevent premature dressing failure due to edge lift Protects the wound and peri-wound skin from contaminants, body fluids, bacteria and viruses
Catalog No. 50-283-1883 Supplier Med Vet International Supplier No. 223931CS
Thin, absorbent and highly conformable May be used as a protective dressing on at-risk skin May be used on intact skin to protect from friction Film border on oval sizes seals to prevent premature dressing failure due to edge lift Protects the wound and peri-wound skin from contaminants, body fluids, bacteria and viruses