Learn More
Med Vet International 3M Tegaderm IV Dressing, Adhesive/Film, 2-3/4 X 3-1/4 inch Sterile, 1633, 100/Box
Catalog No. 502828990
Encompass_Preferred
Transparent film over insertion site allows easy skin and site monitoring Sterile microporous documentation label supports good catheter care practice Two sterile soft cloth tape strips help to anchor catheter and infusion lines Waterproof to protect against external contamination Deep notch design seals around catheter to protect puncture site Design prevents tenting of dressing and stick-to-itself wastage
Catalog No. 50-282-8990 Supplier Med Vet International Supplier No. 331679BX
Transparent film over insertion site allows easy skin and site monitoring Sterile microporous documentation label supports good catheter care practice Two sterile soft cloth tape strips help to anchor catheter and infusion lines Waterproof to protect against external contamination Deep notch design seals around catheter to protect puncture site Design prevents tenting of dressing and stick-to-itself wastage