Learn More
Med Vet International IV Dressing 3M Tegaderm Film 4-3/4 X 4-3/4 inch Sterile, 1668, 25/Box
Catalog No. 502830599
Encompass_Preferred
Adhesive-free window minimizes adhesive contact with needle and skin, reducing the risk of the dressing sticking to needle during removal Flexes with movement and can be worn up to 7 days Transparent dressing permits clear visibility around the insertion site area Transparent film dressing with adhesive free window designed for oncology and hematology patients and others with implanted venous ports
Catalog No. 50-283-0599 Supplier Med Vet International Supplier No. 999387BX
Adhesive-free window minimizes adhesive contact with needle and skin, reducing the risk of the dressing sticking to needle during removal Flexes with movement and can be worn up to 7 days Transparent dressing permits clear visibility around the insertion site area Transparent film dressing with adhesive free window designed for oncology and hematology patients and others with implanted venous ports