Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm Hp 4 X 4-1/2 inch Frame Style DelIVery Oval Sterile, 9546Hp, 200/Case
Catalog No. 502830364
Encompass_Preferred
High adhesion keeps dressings in place under challenging conditions with up to 7-day wear time Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Transparency allows easy monitoring of the wound or I.V. site without the need to remove the dressing Picture-frame delivery makes placement accurate and easy, minimizing potential to stick to gloves or to itself Dressing is conformable and flexes with skin for greater comfort Oval is 4 x 4½ inches
Catalog No. 50-283-0364 Supplier Med Vet International Supplier No. 210039CS
High adhesion keeps dressings in place under challenging conditions with up to 7-day wear time Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Transparency allows easy monitoring of the wound or I.V. site without the need to remove the dressing Picture-frame delivery makes placement accurate and easy, minimizing potential to stick to gloves or to itself Dressing is conformable and flexes with skin for greater comfort Oval is 4 x 4½ inches