Learn More
Med Vet International Foam Dressing 3M Tegaderm 6 X 6-3/4 inch With Border Film Backing Silicone Adhesive Sacral Sterile, 90647, 10/Box
Catalog No. 502830229
Encompass_Preferred
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 6 x 6-3/4 inch sacral shape is good for lower back applications
Catalog No. 50-283-0229 Supplier Med Vet International Supplier No. 1078888BX
Significantly longer wear time while being gentle to the skin Highly conformable Thin, low-profile edge helps to minimize the rolling and lifting that can impact adhesion and wear time Helps maintain the right amount of moisture needed for healing: Multi-layer design absorbs and evaporates moisture to help reduce the potential for skin maceration Easy to apply: Spoke delivery system enables easy application in challenging locations like the sacrum Water-resistant seal means dressing remains intact 6 x 6-3/4 inch sacral shape is good for lower back applications