Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 2 inch X 11 Yard 2 Tab DelIVery Roll Nonsterile, 16002, 4/Case
Catalog No. 502829364
Encompass_Preferred
Provides a waterproof barrier to external contaminants including liquids, bacteria and viruses Can be cut and customized to desired length and shape Can be used for fixation of non-critical medical devices or infusion tubing Dressing is conformable and flexes with skin for greater patient comfort 2 inches x 11 yards
Catalog No. 50-282-9364 Supplier Med Vet International Supplier No. 521718CS
Provides a waterproof barrier to external contaminants including liquids, bacteria and viruses Can be cut and customized to desired length and shape Can be used for fixation of non-critical medical devices or infusion tubing Dressing is conformable and flexes with skin for greater patient comfort 2 inches x 11 yards