Learn More
Med Vet International Dynarex View Guard Transparent Dressings, Sterile, 2 3/8" X 2 3/4", 400/Case
Catalog No. 502832010
Encompass_Preferred
View Guard Transparent dressings protect the skin, IV sites and surgical incisions. Easy to apply Conforms to difficult body contours Provides a breathable barrier that lets air in while keeping moisture vapors and fluid out Ideal for use around IV sites, for wound care and surgical site protection
Catalog No. 50-283-2010 Supplier Med Vet International Supplier No. DYN3642
View Guard Transparent dressings protect the skin, IV sites and surgical incisions. Easy to apply Conforms to difficult body contours Provides a breathable barrier that lets air in while keeping moisture vapors and fluid out Ideal for use around IV sites, for wound care and surgical site protection