Learn More
Med Vet International 3M Tegaderm Transparent Film Dressing, 4 X 4-3/4 inch, Frame Style DelIVery, Rectangle, Sterile, 1626W, 200/Case
Catalog No. 502832446
Encompass_Preferred
3M Tegaderm film creates a sterile and waterproof barrier, sealing out contaminants such as bacteria and fluids It securely holds catheters, feeding tubes and IVs Dressings are transparent for easy monitoring Tegaderm transparent film dressings are also suitable for a wide range of wounds, including pressure ulcers, burns and acute wounds They keep the application area moist for quick healing These dressings are flexible enough to conform to the body and move with the skin 4 x 4-3/4 inches
Catalog No. 50-283-2446 Supplier Med Vet International Supplier No. 121652CS
 Restricted Item
Add to Cart
Add to Cart
3M Tegaderm film creates a sterile and waterproof barrier, sealing out contaminants such as bacteria and fluids It securely holds catheters, feeding tubes and IVs Dressings are transparent for easy monitoring Tegaderm transparent film dressings are also suitable for a wide range of wounds, including pressure ulcers, burns and acute wounds They keep the application area moist for quick healing These dressings are flexible enough to conform to the body and move with the skin 4 x 4-3/4 inches