Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 1-3/4 X 1-3/4 inch Frame Style DelIVery Rectangle Sterile, 1622W, 100/Box
Catalog No. 502829256
Encompass_Preferred
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Adhesive is gentle on skin, and conformable dressing flexes with skin for patient comfort Picture-frame delivery makes accurate placement easy 4.4 x 4.4 centimeters
Catalog No. 50-282-9256 Supplier Med Vet International Supplier No. 264435BX
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Adhesive is gentle on skin, and conformable dressing flexes with skin for patient comfort Picture-frame delivery makes accurate placement easy 4.4 x 4.4 centimeters