Learn More
Med Vet International Foam Dressing Kerrafoam Gentle Border 5 X 5 inch With Border Film Backing Silicone Adhesive Square Sterile, Cwl1013, Each
Catalog No. 502829229
Encompass_Preferred
Soft silicone layer ensures gentle adhesion, excellent absorption rate and nontraumatic removal Smooth outer film layer helps to reduce shear and friction, create a water-resistant bacterial barrier, and maintain an ideal MVTR (moisture vapor transmission rate) Absorbent core and foam pad combine to lock in exudate, bacteria and MMPs – even under compression Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing
Catalog No. 50-282-9229 Supplier Med Vet International Supplier No. 1189361EA
Soft silicone layer ensures gentle adhesion, excellent absorption rate and nontraumatic removal Smooth outer film layer helps to reduce shear and friction, create a water-resistant bacterial barrier, and maintain an ideal MVTR (moisture vapor transmission rate) Absorbent core and foam pad combine to lock in exudate, bacteria and MMPs – even under compression Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing