Learn More
Med Vet International Super Absorbent Dressing Kerramax Care 2 X 2 inch Square, Prd500-025, Each
Catalog No. 502829274
Encompass_Preferred
A super-absorbent dressing that combines a unique wicking layer to distribute exudate throughout the dressing and absorptive polymers that absorb and retain high levels of exudate into a durable yet conformable dressing The unique wicking layer evenly distributes fluid both horizontally and vertically throughout the dressing, utilizing the full absorption capacity Heat-sealed border prevents exudate leakage
Catalog No. 50-282-9274 Supplier Med Vet International Supplier No. 1189375EA
A super-absorbent dressing that combines a unique wicking layer to distribute exudate throughout the dressing and absorptive polymers that absorb and retain high levels of exudate into a durable yet conformable dressing The unique wicking layer evenly distributes fluid both horizontally and vertically throughout the dressing, utilizing the full absorption capacity Heat-sealed border prevents exudate leakage