Learn More
DISCO Hi Tec DRESSER WA600NB50 75 X 75 X 1
Catalog No. NC1928823
Encompass
  • Designed for blade edge shape correction.
  • Optimizes dicing saw blade performance.
  • Utilizes flat dress method for effective conditioning.
  • Removes blade loading from processing materials.
  • Improves processing quality during dicing.
  • Significantly reduces precutassociated costs.
  • Decreases workload related to precutting.
  • Enhances dicing saw operation rates.
  • Length: 75 mm.
  • Width: 75 mm.
  • Thickness: 1 mm.
Catalog No. NC1928823 Supplier DISCO Hi Tec Supplier No. BVDR0003
  • Designed for blade edge shape correction.
  • Optimizes dicing saw blade performance.
  • Utilizes flat dress method for effective conditioning.
  • Removes blade loading from processing materials.
  • Improves processing quality during dicing.
  • Significantly reduces precutassociated costs.
  • Decreases workload related to precutting.
  • Enhances dicing saw operation rates.
  • Length: 75 mm.
  • Width: 75 mm.
  • Thickness: 1 mm.