Learn More
Med Vet International Dynarex Sterile Dynasorb Super Absorbent Dressing, Self-Adhering, 4" X 4", 120/Case
Catalog No. 502829642
Encompass_Preferred
A highly absorbent, multi-layered dressing for use on heavily exuding wounds. Each dressing is soft, comfortable and easy to apply to curved and hard-to-dress areas. Multi-layered dressing with a contact layer and waterproof backing to prevent contamination. Super absorbent polymer core rapidly absorbs exudate, locking it away, minimizing the risk of maceration. Soft and conformable. Pliable enough to allow for a range of movement during use. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing for partial to full thickness at stages 2, 3 or 4. Used to treat leg and pressure ulcers, surgical wounds and donor sites.
Catalog No. 50-282-9642 Supplier Med Vet International Supplier No. 3089
A highly absorbent, multi-layered dressing for use on heavily exuding wounds. Each dressing is soft, comfortable and easy to apply to curved and hard-to-dress areas. Multi-layered dressing with a contact layer and waterproof backing to prevent contamination. Super absorbent polymer core rapidly absorbs exudate, locking it away, minimizing the risk of maceration. Soft and conformable. Pliable enough to allow for a range of movement during use. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing for partial to full thickness at stages 2, 3 or 4. Used to treat leg and pressure ulcers, surgical wounds and donor sites.