Learn More
Med Vet International Absorbent Transparent Film Dressing 3M Tegaderm Absorbent 5-7/8 X 6 inch Rectangle Sterile, 90802, 10/Box
Catalog No. 502832416
Encompass_Preferred
Transparent film and absorbent pad allows wound or surgical incision to be monitored without dressing removal Two tab delivery without label Clear design takes the guesswork out of application over the wound or surgical incision Novel acrylic polymer pad is breathable and designed to absorb low to moderate exudate to promote an optimal healing environment Dressing does not break down in the wound or surgical incision Nonbulky design helps to prevent dressing roll or lift due to friction from linens, clothing or prosthetics Vitro testing demonstrated viral barrier for viruses greater than 27 nanometers in diameter Wear time of up to 7 days gives patients extended protection beyond the initial hospital stay and into the home 5-7/8 x 6 inches
Catalog No. 50-283-2416 Supplier Med Vet International Supplier No. 1214365BX
Transparent film and absorbent pad allows wound or surgical incision to be monitored without dressing removal Two tab delivery without label Clear design takes the guesswork out of application over the wound or surgical incision Novel acrylic polymer pad is breathable and designed to absorb low to moderate exudate to promote an optimal healing environment Dressing does not break down in the wound or surgical incision Nonbulky design helps to prevent dressing roll or lift due to friction from linens, clothing or prosthetics Vitro testing demonstrated viral barrier for viruses greater than 27 nanometers in diameter Wear time of up to 7 days gives patients extended protection beyond the initial hospital stay and into the home 5-7/8 x 6 inches