Learn More
Med Vet International ComfortFoam™ Border Silicone Foam Dressing, 4 x 4 Inch, Sterile, 00317E, 10/Box
Catalog No. 503043166
Encompass_Preferred
ComfortFoam border dressing is self-adherent for easy applications Soft silicone and foam materials make the dressing highly absorbent and flexible Moisture-proof outer film allows for water evaporation, so the dressing stays dry, even after showering Silicone adhesive and a low-profile border hold the dressing in place without sticking to the wound bed, minimizing wound trauma Dressing helps wounds stay moist and thermally insulated for quicker healing
Catalog No. 50-304-3166 Supplier Med Vet International Supplier No. 835617BX
ComfortFoam border dressing is self-adherent for easy applications Soft silicone and foam materials make the dressing highly absorbent and flexible Moisture-proof outer film allows for water evaporation, so the dressing stays dry, even after showering Silicone adhesive and a low-profile border hold the dressing in place without sticking to the wound bed, minimizing wound trauma Dressing helps wounds stay moist and thermally insulated for quicker healing