Learn More
Med Vet International Thin Foam Dressing Mepilex Border Flex Lite 2 X 5 Inch With Border Film Backing Silicone Adhesive Rectangle Sterile, 581100, Each
Catalog No. 503044338
Encompass_Preferred
Four-layer dressing absorbs and channels exudate away from the wound bed and allows you to track exudate progress without disturbing the wound Designed to stay on and conform 2 x 5 inches
Catalog No. 50-304-4338 Supplier Med Vet International Supplier No. 1195995EA
Four-layer dressing absorbs and channels exudate away from the wound bed and allows you to track exudate progress without disturbing the wound Designed to stay on and conform 2 x 5 inches