Learn More
Med Vet International Thin Foam Dressing Mepilex Border Flex Lite 3 X 3 Inch With Border Film Backing Silicone Adhesive Square Sterile, 581200, Each
Catalog No. 503044337
Encompass_Preferred
Mepilex Border Flex Lite is a four-layer, bordered foam dressing that is highly conformable due to Flex Technology It absorbs, channels and traps exudate and allows you to track progress Flex Technology allows MepilexBorder Flex Lite to move in every direction, reducing skin stress, increasing comfort and keeping it in place Mepilex Border Flex may be left in place for up to 7 days depending on the condition of the wound and surrounding skin
Catalog No. 50-304-4337 Supplier Med Vet International Supplier No. 1195996EA
Mepilex Border Flex Lite is a four-layer, bordered foam dressing that is highly conformable due to Flex Technology It absorbs, channels and traps exudate and allows you to track progress Flex Technology allows MepilexBorder Flex Lite to move in every direction, reducing skin stress, increasing comfort and keeping it in place Mepilex Border Flex may be left in place for up to 7 days depending on the condition of the wound and surrounding skin