Learn More
Med Vet International Thin Foam Dressing Mepilex Border Flex Lite 4 X 4 Inch With Border Film Backing Silicone Adhesive Square Sterile, 581300, Each
Catalog No. 503044752
Encompass_Preferred
Four-layer dressing absorbs and channels exudate away from the wound bed and allows you to track exudate progress without disturbing the wound Designed to stay on and conform 4 x 4 inches
Catalog No. 50-304-4752 Supplier Med Vet International Supplier No. 1195997EA
Four-layer dressing absorbs and channels exudate away from the wound bed and allows you to track exudate progress without disturbing the wound Designed to stay on and conform 4 x 4 inches