Learn More
Med Vet International Mepilex Border Flex Foam Dressing, 6 X 8 Inch, W/Border Film Backing, Silicone Adhesive, Rectangle Sterile, 595600, Each
Catalog No. 503043245
Encompass_Preferred
Proprietary Flex technology enables 360° stretch, thanks to patent-pending Y-cuts in the spreading and retention layers Mepilex Border Flex manages both normal and viscous exudate
Catalog No. 50-304-3245 Supplier Med Vet International Supplier No. 1114382EA
Proprietary Flex technology enables 360° stretch, thanks to patent-pending Y-cuts in the spreading and retention layers Mepilex Border Flex manages both normal and viscous exudate