Learn More
Med Vet International Foam Dressing BorderedFoam 4 X 4 Inch With Border Waterproof Backing Nonacrylic Adhesive Square Sterile, 00298E, Each
Catalog No. 503044565
Encompass_Preferred
The 4 x 4 inch foam dressing contains a super absorbent hydrophilic layer to keep the wound area moist It cushions and protects the wound to promote fast healing The strong adhesive backing bonds to the skin creating an airtight seal Waterproof backing reduces the risk of contamination The cushioned foam border insulates, helping to reduce pain
Catalog No. 50-304-4565 Supplier Med Vet International Supplier No. 727739EA
The 4 x 4 inch foam dressing contains a super absorbent hydrophilic layer to keep the wound area moist It cushions and protects the wound to promote fast healing The strong adhesive backing bonds to the skin creating an airtight seal Waterproof backing reduces the risk of contamination The cushioned foam border insulates, helping to reduce pain