Learn More
Med Vet International Lyostypt Collagen Hemostatis Dressing, 3 x 5 cm, J1429, Each
Catalog No. 5000495448
Encompass_Preferred
Lystypt Collagen: fibrillar in thin sheets. Useful for capillary bleeding over large areas, oozing hemorrhages such as spinal surgery. • Can be trimmed for smaller application • Resealable pouch • Absorbed within 3 weeks • Can combine with antibiotics
Catalog No. 50-004-95448 Supplier Med Vet International Supplier No. JOR1429
Lystypt Collagen: fibrillar in thin sheets. Useful for capillary bleeding over large areas, oozing hemorrhages such as spinal surgery. • Can be trimmed for smaller application • Resealable pouch • Absorbed within 3 weeks • Can combine with antibiotics