Learn More
Med Vet International Adhesive Dressing Cosmopor 4 X 4 Inch Nonwoven Square White Sterile, 900820, 25/Box
Catalog No. 503042862
Encompass_Preferred
Soft and gentle to the skin – can be used for those with sensitive skin Conforms well to body contours and remains in place Rounded corners help reduce the need to change the dressing Good absorbency and cushioning effect Fluid repellent backing Sterile
Catalog No. 50-304-2862 Supplier Med Vet International Supplier No. 902132BX
Soft and gentle to the skin – can be used for those with sensitive skin Conforms well to body contours and remains in place Rounded corners help reduce the need to change the dressing Good absorbency and cushioning effect Fluid repellent backing Sterile