Learn More
Med Vet International Foam Dressing DermaFoam 6 X 7 Inch Without Border Waterproof Backing Nonadhesive Elbow / Heel Sterile, 00293E, Each
Catalog No. 503044583
Encompass_Preferred
Dermafoam supports moist wound healing and autolytic debridement Excellent exudate management minimizes maceration Supports normalization of wound bed temperature Absorption, support and cushioning help manage hypergranulation tissue 12.25 x 12.5 centimeters There are five nonadhesive foam dressings in a box
Catalog No. 50-304-4583 Supplier Med Vet International Supplier No. 719720EA
Dermafoam supports moist wound healing and autolytic debridement Excellent exudate management minimizes maceration Supports normalization of wound bed temperature Absorption, support and cushioning help manage hypergranulation tissue 12.25 x 12.5 centimeters There are five nonadhesive foam dressings in a box