Learn More
Med Vet International Foam Dressing DermaLevin 6 X 6 Inch With Border Waterproof Backing Hydrocolloid Adhesive Square Sterile, 00285E, Each
Catalog No. 503043804
Encompass_Preferred
Supports moist wound healing and autolytic debridement Excellent exudate management to minimize maceration Supports normalization of wound bed temperature Conformable for hard to dress areas
Catalog No. 50-304-3804 Supplier Med Vet International Supplier No. 584140EA
Supports moist wound healing and autolytic debridement Excellent exudate management to minimize maceration Supports normalization of wound bed temperature Conformable for hard to dress areas