Learn More
Med Vet International DermaView II™ Transparent Film Dressing with Border, 6½ x 8-3/8 Inch, Sterile, 00254E, Each
Catalog No. 503043582
Encompass_Preferred
Conforms to awkward dressing sites Used to treat superficial wounds, IV sites, burns abrasions and donor sites Waterproof Picture frame border allows for easy application to hard to conform to areas 6½ x 8-3/8 inches
Catalog No. 50-304-3582 Supplier Med Vet International Supplier No. 727083EA
Conforms to awkward dressing sites Used to treat superficial wounds, IV sites, burns abrasions and donor sites Waterproof Picture frame border allows for easy application to hard to conform to areas 6½ x 8-3/8 inches