Learn More
Med Vet International DuoDerm Signal Hydrocolloid Dressing, 4 x 4 Inch, Sterile, 403326, 5/Box
Catalog No. 503044801
Encompass_Preferred
Adhesive hydrocolloid matrix and the outer film on DuoDerm Signal dressing is permeable by moisture vapor Provides an occlusive barrier against bacteria, viruses, dirt and water 10 x 10 centimeters
Catalog No. 50-304-4801 Supplier Med Vet International Supplier No. 426728BX
Adhesive hydrocolloid matrix and the outer film on DuoDerm Signal dressing is permeable by moisture vapor Provides an occlusive barrier against bacteria, viruses, dirt and water 10 x 10 centimeters