Learn More
Med Vet International Curity™ Oil Emulsion Impregnated Dressing, 3 x 8 Inch, Sterile, 6113-, Each
Catalog No. 503045034
Encompass_Preferred
Nonadherent Oil emulsion blend impregnated into open mesh, knitted fabric dressing Non-adhering to wound site Allows free drainage of exudate away from wound Ideal for donor sites, delicate suture lines, graft sites, draining wounds Sterile 3 x 8 inches
Catalog No. 50-304-5034 Supplier Med Vet International Supplier No. 215607EA
Nonadherent Oil emulsion blend impregnated into open mesh, knitted fabric dressing Non-adhering to wound site Allows free drainage of exudate away from wound Ideal for donor sites, delicate suture lines, graft sites, draining wounds Sterile 3 x 8 inches