Learn More
Med Vet International Collagen Dressing Fibracol Plus 2 X 2 inch Square Sterile, 2981, Each
Catalog No. 502830060
Encompass_Preferred
Collagen delivers structural support Alginate provides reliable exudate management
Catalog No. 50-283-0060 Supplier Med Vet International Supplier No. 368787EA
Collagen delivers structural support Alginate provides reliable exudate management