Learn More
Med Vet International Allevyn Adhesive with Border Foam Dressing, 3 x 3 Inch, Sterile, 66020043, 40/Case
Catalog No. 503043070
Encompass_Preferred
Smith & Nephew's Allevyn adhesive foam dressings have three layers for greater fluid absorption and to keep wounds moist to support healing A nonsensitizing adhesive secures dressings without disrupting the wound The top layer is breathable for airflow to help speed healing The flexible material makes them easy to apply and fit around bodily contours These sterile Allevyn wound dressings serve as a protective barrier against bacteria and fluid strikethrough to keep wounds clean They are suitable for many wound types, including burns, diabetic ulcers and shallow injuries 7.5 x 7.5 centimeters
Catalog No. 50-304-3070 Supplier Med Vet International Supplier No. 278016CS
Smith & Nephew's Allevyn adhesive foam dressings have three layers for greater fluid absorption and to keep wounds moist to support healing A nonsensitizing adhesive secures dressings without disrupting the wound The top layer is breathable for airflow to help speed healing The flexible material makes them easy to apply and fit around bodily contours These sterile Allevyn wound dressings serve as a protective barrier against bacteria and fluid strikethrough to keep wounds clean They are suitable for many wound types, including burns, diabetic ulcers and shallow injuries 7.5 x 7.5 centimeters