Learn More
Med Vet International Foam Dressing Allevyn 2 X 2 Inch Without Border Film Backing Nonadhesive Square Sterile, 66027643, 60/Case
Catalog No. 503043649
Encompass_Preferred
Allevyn nonadhesive dressing's absorbent hydrocellular pad, contact layer and breathable top film keep skin moist Dressing creates an environment conducive to healing to help wounds get better faster The outer polyurethane top film is breathable and acts as a barrier to bacteria to prevent infections This 5 x 5 centimeter Allevyn foam dressing has a highly absorbent hydrocellular foam core that wicks away fluids It won't stick to or re-open the wound, ensuring proper healing
Catalog No. 50-304-3649 Supplier Med Vet International Supplier No. 222289CS
Allevyn nonadhesive dressing's absorbent hydrocellular pad, contact layer and breathable top film keep skin moist Dressing creates an environment conducive to healing to help wounds get better faster The outer polyurethane top film is breathable and acts as a barrier to bacteria to prevent infections This 5 x 5 centimeter Allevyn foam dressing has a highly absorbent hydrocellular foam core that wicks away fluids It won't stick to or re-open the wound, ensuring proper healing