Learn More
Sklar Instruments Dressing Forceps, Sklar, 4-1/2 in., Straight, Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501437419 Shop All Sklar Instruments Products
Encompass_Preferred
Dressing Forceps, Sklar, 4-1/2 in., Straight, Serrated, Stainless Steel. Sklar Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. This product is straight with serrated tips and a length of 4-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7419 Supplier Sklar Instruments Supplier No. 191045
Dressing Forceps, Sklar, 4-1/2 in., Straight, Serrated, Stainless Steel. Sklar Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. This product is straight with serrated tips and a length of 4-1/2 inches.