Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Dressing Forceps, Standard Pattern, 4.5IN
SDP

Catalog No. 21520150
Change view
Click to view available options
Length (English):
4.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-520-150 4.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-520-150 Supplier Cenmed™ Supplier No. M91060011
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cenmed™ Dressing Forceps feature serrated handles and tips

Specifications

Autoclavable Autoclavable
Coating Stainless
Color Metallic
Description Tissue Forceps , Standard Pattern, Serrated Handles , 1X2 Teeth , Length: 8
Disposable Reusable
Finish Stainless
For Use With (Application) For grasping tissues in procedures
Length (English) 4.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.