Learn More
Med Vet International Miltex Vantage Dressing Forceps, 5.5in Standard Pattern, Serrated Tips
Catalog No. 502559442
Encompass_Preferred
The Miltex Vantage V96-8 dressing forceps are made from high quality Japanese stainless steel and measure 5.5" in length. They have a non-locking thumb handle and straight, serrated tips.
Catalog No. 50-255-9442 Supplier Med Vet International Supplier No. MILV968
 Restricted Item
Add to Cart
Add to Cart
The Miltex Vantage V96-8 dressing forceps are made from high quality Japanese stainless steel and measure 5.5" in length. They have a non-locking thumb handle and straight, serrated tips.