Learn More
Med Vet International Gelling Fiber Dressing Exufiber 4 X 4-4/5 Inch Rectangle, 709902, Each
Catalog No. 503044044
Encompass_Preferred
Hydrolock technology locks in exudate, blood, and bacteria Viewed under a microscope, you can see that Exufiber is full of tightly entangled fibers that minimize the free space for fluid or exudate to flow, which in turn boosts the entire dressing's integrity
Catalog No. 50-304-4044 Supplier Med Vet International Supplier No. 1158195EA
Hydrolock technology locks in exudate, blood, and bacteria Viewed under a microscope, you can see that Exufiber is full of tightly entangled fibers that minimize the free space for fluid or exudate to flow, which in turn boosts the entire dressing's integrity