Learn More
Med Vet International McKesson Absorbent Gelling Fiber Dressing, 2 x 2 Inch, 87200, Each
Catalog No. 503043208
Encompass_Preferred
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 2 X 2 Inch Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing Low risk of maceration Quilting pattern 2 x 2 inches
Catalog No. 50-304-3208 Supplier Med Vet International Supplier No. 1138625EA
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 2 X 2 Inch Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing Low risk of maceration Quilting pattern 2 x 2 inches