Learn More
Med Vet International McKesson Absorbent Gelling Fiber Dressing, 6 x 6 Inch, 87600, Each
Catalog No. 503044045
Encompass_Preferred
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 6 X 6 Inch Dressings may be used during the healing process of decubitus ulcers (pressure ulcers); venous and arterial leg ulcers; diabetic ulcers; graft and donor sites; post-operative surgical wounds; superficial and partial thickness burns and cavity wounds Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing and low risk of maceration 6 x 6 inches
Catalog No. 50-304-4045 Supplier Med Vet International Supplier No. 1138628EA
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 6 X 6 Inch Dressings may be used during the healing process of decubitus ulcers (pressure ulcers); venous and arterial leg ulcers; diabetic ulcers; graft and donor sites; post-operative surgical wounds; superficial and partial thickness burns and cavity wounds Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing and low risk of maceration 6 x 6 inches