Learn More
Med Vet International Aquacel Extra™ Gelling Fiber Wound Dressing, 6 x 6 Inch, Sterile, 420673, 5/Box
Catalog No. 503044579
Encompass_Preferred
This hydrofiber wound dressing is nine times stronger and 39% more absorbent than a standard Aquacel dressing Aquacel 6 × 6-inch wound dressing is made with two-dimensional strengthening fibers for high strength and durability It gels on contact with fluid for high absorbency It creates a moist environment that helps skin heal faster Use this Aquacel Extra hydrofiber dressing on moderate to highly exuding wounds
Catalog No. 50-304-4579 Supplier Med Vet International Supplier No. 785780BX
This hydrofiber wound dressing is nine times stronger and 39% more absorbent than a standard Aquacel dressing Aquacel 6 × 6-inch wound dressing is made with two-dimensional strengthening fibers for high strength and durability It gels on contact with fluid for high absorbency It creates a moist environment that helps skin heal faster Use this Aquacel Extra hydrofiber dressing on moderate to highly exuding wounds