Learn More
Med Vet International McKesson Nonadhesive without Border Foam Dressing, 4 x 4 Inch, 16-4739, 100/Case
Catalog No. 503044352
Encompass_Preferred
Foam Dressing McKesson 4 X 4 Inch Square Non-Adhesive without Border Sterile Use the hydrocellular foam dressing pads for convenience and efficiency while dressing wounds Works well when covered with a secondary dressing; the pads are designed to absorb medium to heavy fluid discharge White dressing lacks a border to enhance comfort around the sensitive wound site Nonadhesive without border foam dressing is 4 x 4 inches; size can cover small abrasions and wounds
Catalog No. 50-304-4352 Supplier Med Vet International Supplier No. 1138304CS
Foam Dressing McKesson 4 X 4 Inch Square Non-Adhesive without Border Sterile Use the hydrocellular foam dressing pads for convenience and efficiency while dressing wounds Works well when covered with a secondary dressing; the pads are designed to absorb medium to heavy fluid discharge White dressing lacks a border to enhance comfort around the sensitive wound site Nonadhesive without border foam dressing is 4 x 4 inches; size can cover small abrasions and wounds