Learn More
Med Vet International McKesson Nonadhesive without Border Foam Dressing, 4 x 4 Inch, 16-4737, 10/Box
Catalog No. 503044356
Encompass_Preferred
Foam Dressing McKesson 4 X 4 Inch Square Non-Adhesive without Border Sterile Cover with an appropriate secondary dressing depending on the amount of exudate and frequency of dressing change or with medical tape Film backing allows for the dressing to be wiped down and cleaned easily when held in place by tape 4 x 4 inch pad
Catalog No. 50-304-4356 Supplier Med Vet International Supplier No. 1138302BX
Foam Dressing McKesson 4 X 4 Inch Square Non-Adhesive without Border Sterile Cover with an appropriate secondary dressing depending on the amount of exudate and frequency of dressing change or with medical tape Film backing allows for the dressing to be wiped down and cleaned easily when held in place by tape 4 x 4 inch pad