Learn More
Med Vet International McKesson Nonadhesive without Border Foam Dressing, 6 x 6 Inch, 16-4738, 100/Case
Catalog No. 503044335
Encompass_Preferred
Foam Dressing McKesson 6 X 6 Inch Square Non-Adhesive without Border Sterile Helps maintain a moist healing environment at the wound surface Waterproof and breathable outer layer that protects the wound from outside contamination and prevents strike-through of exudate 6 x 6 inch pad
Catalog No. 50-304-4335 Supplier Med Vet International Supplier No. 1138303CS
Foam Dressing McKesson 6 X 6 Inch Square Non-Adhesive without Border Sterile Helps maintain a moist healing environment at the wound surface Waterproof and breathable outer layer that protects the wound from outside contamination and prevents strike-through of exudate 6 x 6 inch pad