Learn More
Med Vet International McKesson Hydrocolloid Dressing, 2 x 2 Inch, 1886, 400/Case
Catalog No. 503045251
Encompass_Preferred
Hydrocolloid Dressing McKesson 2 X 2 Inch Square Sterile The light beige McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a film backing and a highly absorbent inner layer that absorbs wound exudates The sterile dressing is 2 by 2 inches and semi-transparent for visualization The outer layer is moisture resistant to prevent contamination The hydrocolloid dressing has a 7-day wear time
Catalog No. 50-304-5251 Supplier Med Vet International Supplier No. 882991CS
Hydrocolloid Dressing McKesson 2 X 2 Inch Square Sterile The light beige McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a film backing and a highly absorbent inner layer that absorbs wound exudates The sterile dressing is 2 by 2 inches and semi-transparent for visualization The outer layer is moisture resistant to prevent contamination The hydrocolloid dressing has a 7-day wear time