Learn More
Med Vet International Dynarex Island Dressing, Sterile, 4" X 4", 150/Case
Catalog No. 502830848
Encompass_Preferred
Island dressings are non-adherent wound contact layers, individually packed for convenience. Ideal for use as a cover dressing after packing large, deep wounds Individually packed in peel open pouches Provides excellent absorption and cushioning protection Soft, non-stick dressing with adhesive border Not made with natural rubber latex
Catalog No. 50-283-0848 Supplier Med Vet International Supplier No. 3493
Island dressings are non-adherent wound contact layers, individually packed for convenience. Ideal for use as a cover dressing after packing large, deep wounds Individually packed in peel open pouches Provides excellent absorption and cushioning protection Soft, non-stick dressing with adhesive border Not made with natural rubber latex